Anti-AGTRAP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AGTRAP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-733
- 77829-734
- Antigen symbol:
- AGTRAP
- AGTRAP
- Antigen name:
- Type-1 angiotensin II receptor-associated protein (108-152AA)
- Type-1 angiotensin II receptor-associated protein (108-152AA)
- Antigen synonyms:
- Agtrap| Angiotensin II receptor associated protein| Angiotensin II type I receptor associated protein| AT1 receptor-associated protein| ATI receptor associated protein| ATRAP| ATRAP_HUMAN| MGC29646| OTTHUMP00000002269| OTTHUMP00000002270| OTTHUMP00000002271| Type 1 angiotensin II receptor associated protein| Type-1 angiotensin II receptor-associated protein
- Agtrap| Angiotensin II receptor associated protein| Angiotensin II type I receptor associated protein| AT1 receptor-associated protein| ATI receptor associated protein| ATRAP| ATRAP_HUMAN| MGC29646| OTTHUMP00000002269| OTTHUMP00000002270| OTTHUMP00000002271| Type 1 angiotensin II receptor associated protein| Type-1 angiotensin II receptor-associated protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6RW13
- Q6RW13
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together