Anti-SRPK2 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: SRPK2
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77801-038
- 77801-039
- Antigen symbol:
- SRPK2
- SRPK2
- Antigen name:
- SRSF protein kinase 2 protein (471-688AA)
- SRSF protein kinase 2 protein (471-688AA)
- Antigen synonyms:
- Human serine kinase SRPK2 mRNA, complete cds|Serine kinase SRPK2|Serine/arginine rich protein specific kinase 2|Serine/arginine-rich protein-specific kinase 2|Serine/threonine protein kinase SRPK2|SFRS protein kinase 2|SFRSK2|SR protein specific kinase 2|SR-protein-specific kinase 2|SRPK2|SRPK2_HUMAN|SRSF protein kinase 2 C-terminal
- Human serine kinase SRPK2 mRNA, complete cds|Serine kinase SRPK2|Serine/arginine rich protein specific kinase 2|Serine/arginine-rich protein-specific kinase 2|Serine/threonine protein kinase SRPK2|SFRS protein kinase 2|SFRSK2|SR protein specific kinase 2|SR-protein-specific kinase 2|SRPK2|SRPK2_HUMAN|SRSF protein kinase 2 C-terminal
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78362
- P78362
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together