Order Entry
Canada
ContactUsLinkComponent
Anti-METTL2A Rabbit Recombinant Polyclonal Antibody (HRP)
Anti-METTL2A Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-METTL2A Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77834-135
  • 77834-136
  • Antigen symbol:
  • METTL2A
  • METTL2A
  • Antigen name:
  • Methyltransferase-like protein 2A protein (1-242AA)
  • Methyltransferase-like protein 2A protein (1-242AA)
  • Antigen synonyms:
  • METTL2A|METTL2|HSPC266|tRNA N(3)-methylcytidine methyltransferase METTL2A|EC 2.1.1.-|Methyltransferase-like protein 2A
  • METTL2A|METTL2|HSPC266|tRNA N(3)-methylcytidine methyltransferase METTL2A|EC 2.1.1.-|Methyltransferase-like protein 2A
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q96IZ6
  • Q96IZ6
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together