Anti-FAIM2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: FAIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77813-930
- 77813-931
- Antigen symbol:
- FAIM2
- FAIM2
- Antigen name:
- Fusion protein of FAIM2
- Fusion protein of FAIM2
- Antigen synonyms:
- FAIM 2|Faim2|FAIM2_HUMAN|Fas apoptotic inhibitory molecule 2|KIAA0950|LFG|Lifeguard|Neural membrane protein 35|NGP 35|NGP35|NMP 35|NMP35|Protein lifeguard|TMBIM 2|TMBIM2|Transmembrane BAX inhibitor motif containing 2|Transmembrane BAX inhibitor motif containing protein 2|Transmembrane BAX inhibitor motif-containing protein 2
- FAIM 2|Faim2|FAIM2_HUMAN|Fas apoptotic inhibitory molecule 2|KIAA0950|LFG|Lifeguard|Neural membrane protein 35|NGP 35|NGP35|NMP 35|NMP35|Protein lifeguard|TMBIM 2|TMBIM2|Transmembrane BAX inhibitor motif containing 2|Transmembrane BAX inhibitor motif containing protein 2|Transmembrane BAX inhibitor motif-containing protein 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BWQ8
- Q9BWQ8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together