Anti-SLCO1C1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLCO1C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-593
- 77837-592
- Antigen symbol:
- SLCO1C1
- SLCO1C1
- Antigen name:
- Solute carrier organic anion transporter family member 1C1 protein (297-348AA)
- Solute carrier organic anion transporter family member 1C1 protein (297-348AA)
- Antigen synonyms:
- OAT-RP-5|OATP-14|OATP-F|OATP1|OATP1C1|OATPRP5|Organic anion transporter F|Organic anion transporter polypeptide-related protein 5|organic anion transporting polypeptide 14|Organic anion-transporting polypeptide 14|SLC21A14|Slco1c1|SO1C1_HUMAN|solute carrier family 21 (organic anion transporter), member 14|Solute carrier family 21 member 14|Solute carrier organic anion transporter family member 1C1|solute carrier organic anion transporter family, member 1C1|Thyroxine transporter
- OAT-RP-5|OATP-14|OATP-F|OATP1|OATP1C1|OATPRP5|Organic anion transporter F|Organic anion transporter polypeptide-related protein 5|organic anion transporting polypeptide 14|Organic anion-transporting polypeptide 14|SLC21A14|Slco1c1|SO1C1_HUMAN|solute carrier family 21 (organic anion transporter), member 14|Solute carrier family 21 member 14|Solute carrier organic anion transporter family member 1C1|solute carrier organic anion transporter family, member 1C1|Thyroxine transporter
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NYB5
- Q9NYB5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together