Anti-DDX59 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DDX59
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-404
- 77828-405
- Antigen symbol:
- DDX59
- DDX59
- Antigen name:
- Probable ATP-dependent RNA helicase DDX59 protein (1-200AA)
- Probable ATP-dependent RNA helicase DDX59 protein (1-200AA)
- Antigen synonyms:
- DDX 59|DDX59|DDX59_HUMAN|DEAD (Asp-Glu-Ala-Asp) box polypeptide 59|DEAD box protein 59|DKFZP564B1023|OTTHUMP00000033840|OTTHUMP00000033841|Probable ATP-dependent RNA helicase DDX59|RP11-92G12.2|Zinc finger HIT domain containing protein 5|Zinc finger HIT domain-containing protein 5|ZNHIT5
- DDX 59|DDX59|DDX59_HUMAN|DEAD (Asp-Glu-Ala-Asp) box polypeptide 59|DEAD box protein 59|DKFZP564B1023|OTTHUMP00000033840|OTTHUMP00000033841|Probable ATP-dependent RNA helicase DDX59|RP11-92G12.2|Zinc finger HIT domain containing protein 5|Zinc finger HIT domain-containing protein 5|ZNHIT5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5T1V6
- Q5T1V6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together