Anti-DSC3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DSC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-027
- 77822-026
- Antigen symbol:
- DSC3
- DSC3
- Antigen name:
- Desmocollin-3 protein (712-896AA)
- Desmocollin-3 protein (712-896AA)
- Antigen synonyms:
- Cadherin family member 3|CDHF3|desmocollin 3a/3b|Desmocollin 4|Desmocollin-3|Desmocollin-4|Desmocollin3|Desmocollin4|DSC|DSC1|DSC2|DSC3|DSC3_HUMAN|DSC4|HT CP|HT-CP|HTCP
- Cadherin family member 3|CDHF3|desmocollin 3a/3b|Desmocollin 4|Desmocollin-3|Desmocollin-4|Desmocollin3|Desmocollin4|DSC|DSC1|DSC2|DSC3|DSC3_HUMAN|DSC4|HT CP|HT-CP|HTCP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14574
- Q14574
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together