Anti-SH3BP5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SH3BP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-485
- 77799-486
- Antigen symbol:
- SH3BP5
- SH3BP5
- Antigen name:
- SH3 domain-binding protein 5 protein (49-455AA)
- SH3 domain-binding protein 5 protein (49-455AA)
- Antigen synonyms:
- 3BP5_HUMAN|OTTHUMP00000208289|OTTHUMP00000208291|OTTHUMP00000208294|SAB|SH3 binding protein|SH3 domain binding protein 5 (BTK associated)|SH3 domain binding protein 5|SH3 domain binding protein that preferentially associates with BTK|SH3 domain-binding protein 5|SH3 domain-binding protein that preferentially associates with BTK|SH3 domainbinding protein 5|SH3BP 5|SH3BP-5|Sh3bp5
- 3BP5_HUMAN|OTTHUMP00000208289|OTTHUMP00000208291|OTTHUMP00000208294|SAB|SH3 binding protein|SH3 domain binding protein 5 (BTK associated)|SH3 domain binding protein 5|SH3 domain binding protein that preferentially associates with BTK|SH3 domain-binding protein 5|SH3 domain-binding protein that preferentially associates with BTK|SH3 domainbinding protein 5|SH3BP 5|SH3BP-5|Sh3bp5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60239
- O60239
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together