Anti-AVPR1B Rabbit Polyclonal Antibody
Catalog # 77826-055
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthetic peptide from protein at AA range: 271-320
- Antigen symbol:AVPR1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P47901
- Antigen synonyms:Antidiuretic hormone receptor 1b|Antidiuretic hormone|receptor for|V1B|Arginine vasopressin receptor 1B|Arginine vasopressin receptor 3|AVPR V1b|AVPR V3|Avpr1b|AVPR3|Pituitary vasopressin receptor 3|V1bR|V1BR_HUMAN|Vasopressin V1b receptor|Vasopressin V3 receptor|VPR3
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 300, ELISA: 1:10000 to 20000
Type:
Antigen: AVPR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat