Anti-CHAF1A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: CHAF1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-379
- 77823-378
- Antigen symbol:
- CHAF1A
- CHAF1A
- Antigen name:
- Chromatin assembly factor 1 subunit A protein (840-904AA)
- Chromatin assembly factor 1 subunit A protein (840-904AA)
- Antigen synonyms:
- CAF 1| CAF 1 subunit A| CAF| CAF I 150 kDa subunit| CAF I p150| CAF-1 subunit A| CAF-I 150 kDa subunit| CAF-I p150| CAF1| CAF1A_HUMAN| CAF1B| CAF1P150| CHAF1A| Chromatin assembly factor 1 subunit A| Chromatin Assembly Factor 1 Subunit A p150| Chromatin assembly factor I (150 kDa)| Chromatin assembly factor I 150 kDa| Chromatin assembly factor I p150 subunit| DCAF1| hp150| MGC71229| P150
- CAF 1| CAF 1 subunit A| CAF| CAF I 150 kDa subunit| CAF I p150| CAF-1 subunit A| CAF-I 150 kDa subunit| CAF-I p150| CAF1| CAF1A_HUMAN| CAF1B| CAF1P150| CHAF1A| Chromatin assembly factor 1 subunit A| Chromatin Assembly Factor 1 Subunit A p150| Chromatin assembly factor I (150 kDa)| Chromatin assembly factor I 150 kDa| Chromatin assembly factor I p150 subunit| DCAF1| hp150| MGC71229| P150
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13111
- Q13111
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together