Anti-Reg3b Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Reg3b
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Specifications
- Catalog No:
- 77815-702
- 77815-701
- Antigen symbol:
- Reg3b
- Reg3b
- Antigen name:
- Recombinant Rat Regenerating islet-derived protein 3-beta protein (27-175AA)
- Recombinant Rat Regenerating islet-derived protein 3-beta protein (27-175AA)
- Antigen synonyms:
- Reg3b|Pap|Pap1|Reg2|Regenerating islet-derived protein 3-beta|REG-3-beta|Pancreatitis-associated protein 1|Peptide 23|REG-2|Regenerating islet-derived protein III-beta|Reg III-beta) [Cleaved into: Regenerating islet-derived protein 3-beta 16.5 kDa form|Regenerating islet-derived protein 3-beta 15 kDa form]
- Reg3b|Pap|Pap1|Reg2|Regenerating islet-derived protein 3-beta|REG-3-beta|Pancreatitis-associated protein 1|Peptide 23|REG-2|Regenerating islet-derived protein III-beta|Reg III-beta) [Cleaved into: Regenerating islet-derived protein 3-beta 16.5 kDa form|Regenerating islet-derived protein 3-beta 15 kDa form]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Rat
- Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25031
- P25031
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together