Anti-PKD2 Rabbit Polyclonal Antibody
77806-186
New Product
- Antigen name:Synthesized peptide derived from PKD2 around the non-phosphorylation site of S812.
- Antigen symbol:PKD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q13563
- Antigen synonyms:APKD2|Autosomal dominant polycystic kidney disease type II|Autosomal dominant polycystic kidney disease type II protein|MGC138466|MGC138468|PC 2|PC2|PKD 2|PKD2|PKD2_HUMAN|PKD4|Polycystic kidney disease 2 (autosomal dominant)|Polycystic kidney disease 2|Polycystic kidney disease 2 protein|Polycystin 2|Polycystin 2 transient receptor potential cation channel|Polycystin-2|Polycystin2|Polycystwin|R48321|Transient receptor potential cation channel subfamily P member 2|TRPP2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IF: 1:200 to 1:1000, ELISA: 1:10000
Type:
Antigen: PKD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat