Anti-SLC23A3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC23A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-157
- 77831-156
- Antigen symbol:
- SLC23A3
- SLC23A3
- Antigen name:
- Solute carrier family 23 member 3 protein (391-492AA)
- Solute carrier family 23 member 3 protein (391-492AA)
- Antigen synonyms:
- E2 binding protein 3|E2BP3|FLJ31168|HPC E2 binding protein 3|HPC E2-binding protein 3|Na(+)/ Lascorbic acid transporter 3|Na(+)/L-ascorbic acid transporter 3|S23A3|S23A3_HUMAN|Slc23a3|Sodium dependent vitamin C transporter 3|Sodium-dependent vitamin C transporter 3|Solute carrier family 23 member 3|Solute carrier family 23 nucleobase transporters member 3|SVCT3|Yspl1
- E2 binding protein 3|E2BP3|FLJ31168|HPC E2 binding protein 3|HPC E2-binding protein 3|Na(+)/ Lascorbic acid transporter 3|Na(+)/L-ascorbic acid transporter 3|S23A3|S23A3_HUMAN|Slc23a3|Sodium dependent vitamin C transporter 3|Sodium-dependent vitamin C transporter 3|Solute carrier family 23 member 3|Solute carrier family 23 nucleobase transporters member 3|SVCT3|Yspl1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6PIS1
- Q6PIS1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together