Anti-CSF3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CSF3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77809-776
- 77809-775
- Antigen symbol:
- CSF3
- CSF3
- Antigen name:
- Granulocyte colony-stimulating factor protein (27-200AA)
- Granulocyte colony-stimulating factor protein (27-200AA)
- Antigen synonyms:
- C17orf33| Colony stimulating factor 3 (granulocyte)| Colony stimulating factor 3| CSF 3| CSF beta| CSF3| CSF3_HUMAN| CSF3OS| Csfg| Filgrastim| G-CSF| GCSA| GCSF| Granulocyte colony stimulating factor| Granulocyte colony-stimulating factor| Lenograstim| Macrophage granulocyte inducer 2| MGC45931| MGI 2| Pluripoietin
- C17orf33| Colony stimulating factor 3 (granulocyte)| Colony stimulating factor 3| CSF 3| CSF beta| CSF3| CSF3_HUMAN| CSF3OS| Csfg| Filgrastim| G-CSF| GCSA| GCSF| Granulocyte colony stimulating factor| Granulocyte colony-stimulating factor| Lenograstim| Macrophage granulocyte inducer 2| MGC45931| MGI 2| Pluripoietin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09919
- P09919
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together