Anti-POMP Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: POMP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-818
- 77794-819
- Antigen symbol:
- POMP
- POMP
- Antigen name:
- Proteasome maturation protein (1-141AA)
- Proteasome maturation protein (1-141AA)
- Antigen synonyms:
- 2510048O06Rik|C13orf12|Chromosome 13 open reading frame 12|HSPC 014|HSPC036 protein|hUMP 1|hUMP1|PNAS 110|PNAS110|Pomp|POMP_HUMAN|Proteasome maturation protein|Proteassemblin|Protein UMP1 homolog|UMP 1|UMP1|yeast|homolog of|Voltage gated K channel beta subunit 4.1|Voltage-gated K channel beta subunit 4.1|voltage-gated potassium channel beta subunit 4.1
- 2510048O06Rik|C13orf12|Chromosome 13 open reading frame 12|HSPC 014|HSPC036 protein|hUMP 1|hUMP1|PNAS 110|PNAS110|Pomp|POMP_HUMAN|Proteasome maturation protein|Proteassemblin|Protein UMP1 homolog|UMP 1|UMP1|yeast|homolog of|Voltage gated K channel beta subunit 4.1|Voltage-gated K channel beta subunit 4.1|voltage-gated potassium channel beta subunit 4.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y244
- Q9Y244
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together