Anti-FKBP14 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: FKBP14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77836-576
- 77836-575
- Antigen symbol:
- FKBP14
- FKBP14
- Antigen name:
- Peptidyl-prolyl cis-trans isomerase FKBP14 protein (20-211AA)
- Peptidyl-prolyl cis-trans isomerase FKBP14 protein (20-211AA)
- Antigen synonyms:
- 22 kDa FK506 binding protein|22 kDa FK506-binding protein|22 kDa FKBP|FK506 binding protein 14 (22 kDa)|FK506 binding protein 14|FK506-binding protein 14|FKB14_HUMAN|FKBP 22|FKBP-14|FKBP-22|FKBP14|FKBP22|FLJ20731|Peptidyl prolyl cis trans isomerase|Peptidyl-prolyl cis-trans isomerase FKBP14|PPIase|PPIase FKBP14|Rotamase
- 22 kDa FK506 binding protein|22 kDa FK506-binding protein|22 kDa FKBP|FK506 binding protein 14 (22 kDa)|FK506 binding protein 14|FK506-binding protein 14|FKB14_HUMAN|FKBP 22|FKBP-14|FKBP-22|FKBP14|FKBP22|FLJ20731|Peptidyl prolyl cis trans isomerase|Peptidyl-prolyl cis-trans isomerase FKBP14|PPIase|PPIase FKBP14|Rotamase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NWM8
- Q9NWM8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together