Anti-PPCDC Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPCDC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-943
- 77794-944
- Antigen symbol:
- PPCDC
- PPCDC
- Antigen name:
- Phosphopantothenoylcysteine decarboxylase protein (51-183AA)
- Phosphopantothenoylcysteine decarboxylase protein (51-183AA)
- Antigen synonyms:
- 1810057I13Rik|8430432M10Rik|CoaC|COAC_HUMAN|FLJ14585|MDS018|MGC102190|Phosphopantothenoylcysteine decarboxylase|PPC-DC|ppcdc|RGD1306267
- 1810057I13Rik|8430432M10Rik|CoaC|COAC_HUMAN|FLJ14585|MDS018|MGC102190|Phosphopantothenoylcysteine decarboxylase|PPC-DC|ppcdc|RGD1306267
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96CD2
- Q96CD2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together