Anti-TRIM8 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:200 to 1:500, IF:1:50 to 1:200
Type:
Antigen: TRIM8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-174
- 77838-173
- Antigen symbol:
- TRIM8
- TRIM8
- Antigen name:
- Probable E3 ubiquitin-protein ligase TRIM8 protein (367-517AA)
- Probable E3 ubiquitin-protein ligase TRIM8 protein (367-517AA)
- Antigen synonyms:
- AA408830|BB006755|GERP|Glioblastoma-expressed RING finger protein|Lioblastoma expressed ring finger protein|Probable E3 ubiquitin-protein ligase TRIM8|RING finger protein 27|RNF27|RP11-47A8.4|TRIM8|TRIM8_HUMAN|Tripartite motif containing 8|Tripartite motif protein TRIM8|Tripartite motif-containing protein 8
- AA408830|BB006755|GERP|Glioblastoma-expressed RING finger protein|Lioblastoma expressed ring finger protein|Probable E3 ubiquitin-protein ligase TRIM8|RING finger protein 27|RNF27|RP11-47A8.4|TRIM8|TRIM8_HUMAN|Tripartite motif containing 8|Tripartite motif protein TRIM8|Tripartite motif-containing protein 8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BZR9
- Q9BZR9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together