Anti-cssB Rabbit Recombinant Polyclonal Antibody (FITC)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: cssB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Spécifications
- Catalog No:
- 77816-601
- 77816-602
- Antigen symbol:
- cssB
- cssB
- Antigen name:
- Recombinant Escherichia coli CS6 fimbrial subunit B protein (22-167AA)
- Recombinant Escherichia coli CS6 fimbrial subunit B protein (22-167AA)
- Antigen synonyms:
- cssBCS6 fimbrial subunit B
- cssBCS6 fimbrial subunit B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- E. coli
- E. coli
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53510
- P53510
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Spécifications
Frequently Bought Together