Anti-TGFB2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TGFB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77810-802
- 77810-801
- Antigen symbol:
- TGFB2
- TGFB2
- Antigen name:
- Transforming growth factor beta-2 proprotein protein (304-413AA)
- Transforming growth factor beta-2 proprotein protein (304-413AA)
- Antigen synonyms:
- BSC-1 cell growth inhibitor|BSC1 cell growth inhibitor|Cetermin|G-TSF|Glioblastoma-derived T-cell suppressor factor|GTSF|LAP|Latency-associated peptide|MGC116892|MGF|Milk growth factor|Polyergin|TGF-beta-2|TGF-beta2|TGFB2|TGFB2_HUMAN|Transforming growth factor beta 2
- BSC-1 cell growth inhibitor|BSC1 cell growth inhibitor|Cetermin|G-TSF|Glioblastoma-derived T-cell suppressor factor|GTSF|LAP|Latency-associated peptide|MGC116892|MGF|Milk growth factor|Polyergin|TGF-beta-2|TGF-beta2|TGFB2|TGFB2_HUMAN|Transforming growth factor beta 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61812
- P61812
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together