Anti-POLR2C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POLR2C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-737
- 77794-738
- Antigen symbol:
- POLR2C
- POLR2C
- Antigen name:
- DNA-directed RNA polymerase II subunit RPB3 protein (2-275AA)
- DNA-directed RNA polymerase II subunit RPB3 protein (2-275AA)
- Antigen synonyms:
- DNA directed RNA polymerase II 33 kDa polypeptide|DNA directed RNA polymerase II subunit C|DNA directed RNA polymerase II subunit RPB3|DNA-directed RNA polymerase II 33 kDa polypeptide|DNA-directed RNA polymerase II subunit C|DNA-directed RNA polymerase II subunit RPB3|hRPB33|hsRPB3|POLR 2C|POLR2 C|Polr2c|Polymerase (RNA) II (DNA directed) polypeptide C|33kDa|polymerase RNA II DNA directed polypeptide C 33kDa|RNA polymerase II polypeptide C|RNA polymerase II subunit 3|RNA polymerase II subunit B3|RPB 3|RPB3|RPB3_HUMAN|RPB31|RPB33
- DNA directed RNA polymerase II 33 kDa polypeptide|DNA directed RNA polymerase II subunit C|DNA directed RNA polymerase II subunit RPB3|DNA-directed RNA polymerase II 33 kDa polypeptide|DNA-directed RNA polymerase II subunit C|DNA-directed RNA polymerase II subunit RPB3|hRPB33|hsRPB3|POLR 2C|POLR2 C|Polr2c|Polymerase (RNA) II (DNA directed) polypeptide C|33kDa|polymerase RNA II DNA directed polypeptide C 33kDa|RNA polymerase II polypeptide C|RNA polymerase II subunit 3|RNA polymerase II subunit B3|RPB 3|RPB3|RPB3_HUMAN|RPB31|RPB33
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19387
- P19387
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together