Anti-RHBDL2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77797-260
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Rhomboid-related protein 2 protein (12-132AA)
- Antigen symbol:RHBDL2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:Q9NX52
- Antigen synonyms:RHBDL2| Rhomboid-related protein 2| RRP2| EC 3.4.21.105| Rhomboid-like protein 2) [Cleaved into: Rhomboid-related protein 2| N-terminal fragment| NTF)| C-terminal fragment| CTF)]
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RHBDL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human