Anti-RB1CC1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: RB1CC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-592
- 77833-593
- Antigen symbol:
- RB1CC1
- RB1CC1
- Antigen name:
- RB1-inducible coiled-coil protein 1 protein (1544-1594AA)
- RB1-inducible coiled-coil protein 1 protein (1544-1594AA)
- Antigen synonyms:
- FAK family kinase-interacting protein of 200 kDa|FIP200|RB1-inducible coiled-coil protein 1||RBCC1_HUMAN
- FAK family kinase-interacting protein of 200 kDa|FIP200|RB1-inducible coiled-coil protein 1||RBCC1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDY2
- Q8TDY2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together