Anti-PRPSAP1 Rabbit Polyclonal Antibody
Catalog # 77795-604
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:39 kDa phosphoribosypyrophosphate synthase associated protein
- Antigen symbol:PRPSAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q14558
- Antigen synonyms:39 kDa phosphoribosypyrophosphate synthase associated protein|39 kDa phosphoribosypyrophosphate synthase-associated protein|KPRA_HUMAN|PAP39|Phosphoribosyl pyrophosphate synthase associated protein 1|Phosphoribosyl pyrophosphate synthase-associated protein 1|Phosphoribosyl pyrophosphate synthetase associated protein 1|PRPP synthase associated protein 1|PRPP synthase-associated protein 1|Prpsap1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PRPSAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat