Anti-PLEKHA8 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLEKHA8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-143
- 77834-142
- Antigen symbol:
- PLEKHA8
- PLEKHA8
- Antigen name:
- Pleckstrin homology domain-containing family A member 8 protein (1-300AA)
- Pleckstrin homology domain-containing family A member 8 protein (1-300AA)
- Antigen synonyms:
- FAPP-2|FAPP2|hFAPP2|MGC3358|PH domain-containing family A member 8|Phosphatidylinositol-four-phosphate adapter protein 2|Phosphoinositide binding specificmember 8|Phosphoinositol 4 phosphate adaptor protein 2|Phosphoinositol 4-phosphate adapter protein 2|PKHA8_HUMAN|Pleckstrin homology domain containing A8|Pleckstrin homology domain containing family A (Phosphoinositide binding specific) member 8|Pleckstrin homology domain containing family A|Pleckstrin homology domain-containing family A member 8|Plekha8|PLEKHA8 protein|Serologically defined breast cancer antigen NY-BR-86
- FAPP-2|FAPP2|hFAPP2|MGC3358|PH domain-containing family A member 8|Phosphatidylinositol-four-phosphate adapter protein 2|Phosphoinositide binding specificmember 8|Phosphoinositol 4 phosphate adaptor protein 2|Phosphoinositol 4-phosphate adapter protein 2|PKHA8_HUMAN|Pleckstrin homology domain containing A8|Pleckstrin homology domain containing family A (Phosphoinositide binding specific) member 8|Pleckstrin homology domain containing family A|Pleckstrin homology domain-containing family A member 8|Plekha8|PLEKHA8 protein|Serologically defined breast cancer antigen NY-BR-86
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96JA3
- Q96JA3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together