Anti-HBEGF Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HBEGF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-380
- 77834-379
- Antigen symbol:
- HBEGF
- HBEGF
- Antigen name:
- Proheparin-binding EGF-like growth factor protein (63-148AA)
- Proheparin-binding EGF-like growth factor protein (63-148AA)
- Antigen synonyms:
- diphtheria toxin receptor (heparin-binding EGF-like growth factor)| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growth factor)| Diphtheria toxin receptor| DT R| DT-R| DTR| DTS| DTSF| HB-EGF| HBEGF| HBEGF_HUMAN| HEGFL| Heparin binding EGF like growth factor| Heparin binding epidermal growth factor| Heparin binding epidermal growth factor like growth factor| Heparin-binding EGF-like growth factor| Proheparin binding EGF like growth factor
- diphtheria toxin receptor (heparin-binding EGF-like growth factor)| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growth factor)| Diphtheria toxin receptor| DT R| DT-R| DTR| DTS| DTSF| HB-EGF| HBEGF| HBEGF_HUMAN| HEGFL| Heparin binding EGF like growth factor| Heparin binding epidermal growth factor| Heparin binding epidermal growth factor like growth factor| Heparin-binding EGF-like growth factor| Proheparin binding EGF like growth factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99075
- Q99075
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together