Anti-PG_1424 Rabbit Recombinant Polyclonal Antibody
77842-629
New Product
- Antigen name:Recombinant Porphyromonas gingivalis Peptidylarginine deiminase protein (44-556AA)
- Antigen symbol:PG_1424
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Porphyromonas gingivalis
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9RQJ2
- Antigen synonyms:PG_1424Peptidylarginine deiminase|EC 3.5.3.-
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: PG_1424
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Porphyromonas gingivalis