Anti-PTH2R Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PTH2R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-041
- 77796-040
- Antigen symbol:
- PTH2R
- PTH2R
- Antigen name:
- Parathyroid hormone 2 receptor protein (27-145AA)
- Parathyroid hormone 2 receptor protein (27-145AA)
- Antigen synonyms:
- Parathyroid hormone 2 receptor|Parathyroid hormone receptor precursor|PTH 2 receptor|PTH2 receptor|Pth2r|PTH2R_HUMAN|Pthr 2|Pthr2
- Parathyroid hormone 2 receptor|Parathyroid hormone receptor precursor|PTH 2 receptor|PTH2 receptor|Pth2r|PTH2R_HUMAN|Pthr 2|Pthr2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49190
- P49190
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together