Anti-PAG1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PAG1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-481
- 77842-480
- Antigen symbol:
- PAG1
- PAG1
- Antigen name:
- Phosphoprotein associated with glycosphingolipid-enriched microdomains 1 protein (97-432AA)
- Phosphoprotein associated with glycosphingolipid-enriched microdomains 1 protein (97-432AA)
- Antigen synonyms:
- CBP|Csk binding protein|Csk-binding protein|FLJ37858|MGC138364|PAG 1|PAG|PAG1|PAG1_HUMAN|Phosphoprotein associated with glycosphingolipid enriched microdomains 1|Phosphoprotein associated with glycosphingolipid enriched microdomains|Phosphoprotein associated with glycosphingolipid microdomains 1|Phosphoprotein associated with glycosphingolipid-enriched microdomains 1|Protein Associated with Glycosphingolipid Enriched Microdomains|Transmembrane adapter protein PAG|Transmembrane phosphoprotein Cbp
- CBP|Csk binding protein|Csk-binding protein|FLJ37858|MGC138364|PAG 1|PAG|PAG1|PAG1_HUMAN|Phosphoprotein associated with glycosphingolipid enriched microdomains 1|Phosphoprotein associated with glycosphingolipid enriched microdomains|Phosphoprotein associated with glycosphingolipid microdomains 1|Phosphoprotein associated with glycosphingolipid-enriched microdomains 1|Protein Associated with Glycosphingolipid Enriched Microdomains|Transmembrane adapter protein PAG|Transmembrane phosphoprotein Cbp
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NWQ8
- Q9NWQ8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together