Anti-FAM118A Rabbit Recombinant Polyclonal Antibody
77842-482
New Product
- Antigen name:Protein FAM118A protein (51-357AA)
- Antigen symbol:FAM118A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NWS6
- Antigen synonyms:bK268H5.C22.4|C22orf8|F118A_HUMAN|Fam118a|Family with sequence similarity 118|member A|Hypothetical protein LOC55007|Protein FAM118A
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: FAM118A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human