Anti-SLC34A3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC34A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-280
- 77833-279
- Antigen symbol:
- SLC34A3
- SLC34A3
- Antigen name:
- Sodium-dependent phosphate transport protein 2C protein (1-76AA)
- Sodium-dependent phosphate transport protein 2C protein (1-76AA)
- Antigen synonyms:
- HHRH | Na(+) dependent phosphate cotransporter 2C | Na(+) Pi cotransporter 2C| Na(+)-dependent phosphate cotransporter 2C| Na(+)/Pi cotransporter 2C| NaPi 2c| NaPi-2c| NPT2C | NPT2C_HUMAN| NPTIIC | SLC34A3| Sodium dependent phosphate transport protein 2C| Sodium inorganic phosphate cotransporter IIC| Sodium phosphate cotransporter 2C| Sodium phosphate transport protein 2C | Sodium-dependent phosphate transport protein 2C| Sodium-phosphate transport protein 2C| Sodium/inorganic phosphate cotransporter IIC| Sodium/inorganic phosphate cotransporter| type IIC| Sodium/phosphate cotransporter 2C| solute carrier family 34 (sodium phosphate) member 3| solute carrier family 34 (sodium/phosphate contransporter)| member 3| solute carrier family 34 (type II sodium/phosphate contransporter)| Solute carrier family 34 member 3| type IIc Na+ Pi cotransporter | Type IIc Na+/Pi cotransporter
- HHRH | Na(+) dependent phosphate cotransporter 2C | Na(+) Pi cotransporter 2C| Na(+)-dependent phosphate cotransporter 2C| Na(+)/Pi cotransporter 2C| NaPi 2c| NaPi-2c| NPT2C | NPT2C_HUMAN| NPTIIC | SLC34A3| Sodium dependent phosphate transport protein 2C| Sodium inorganic phosphate cotransporter IIC| Sodium phosphate cotransporter 2C| Sodium phosphate transport protein 2C | Sodium-dependent phosphate transport protein 2C| Sodium-phosphate transport protein 2C| Sodium/inorganic phosphate cotransporter IIC| Sodium/inorganic phosphate cotransporter| type IIC| Sodium/phosphate cotransporter 2C| solute carrier family 34 (sodium phosphate) member 3| solute carrier family 34 (sodium/phosphate contransporter)| member 3| solute carrier family 34 (type II sodium/phosphate contransporter)| Solute carrier family 34 member 3| type IIc Na+ Pi cotransporter | Type IIc Na+/Pi cotransporter
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N130
- Q8N130
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together