Anti-EIF3I Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EIF3I
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-497
- 77806-496
- Antigen symbol:
- EIF3I
- EIF3I
- Antigen name:
- Eukaryotic translation initiation factor 3 subunit I protein (1-323AA)
- Eukaryotic translation initiation factor 3 subunit I protein (1-323AA)
- Antigen synonyms:
- eIF-3-beta|eIF3 beta|eIF3 p36|eIF3-beta|eIF3-p36|eIF3i|EIF3I_HUMAN|EIF3S2|formerly|Eukaryotic translation initiation factor 3 subunit 2|Eukaryotic translation initiation factor 3 subunit 2 beta|Eukaryotic translation initiation factor 3 subunit I|eukaryotic translation initiation factor 3|subunit 2 (beta|36kD|subunit 2 beta|36kDa|subunit 2|subunit I|Predicted protein of HQ2242|PRO2242|TGF beta receptor interacting protein 1|TGF-beta receptor-interacting protein 1|TGFbeta receptor interacting protein 1|TGFbeta receptor-interacting protein 1|TRIP 1|TRIP-1|TRIP1
- eIF-3-beta|eIF3 beta|eIF3 p36|eIF3-beta|eIF3-p36|eIF3i|EIF3I_HUMAN|EIF3S2|formerly|Eukaryotic translation initiation factor 3 subunit 2|Eukaryotic translation initiation factor 3 subunit 2 beta|Eukaryotic translation initiation factor 3 subunit I|eukaryotic translation initiation factor 3|subunit 2 (beta|36kD|subunit 2 beta|36kDa|subunit 2|subunit I|Predicted protein of HQ2242|PRO2242|TGF beta receptor interacting protein 1|TGF-beta receptor-interacting protein 1|TGFbeta receptor interacting protein 1|TGFbeta receptor-interacting protein 1|TRIP 1|TRIP-1|TRIP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13347
- Q13347
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together