Anti-WIPI2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77819-617
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Recombinant Protein
- Antigen symbol:WIPI2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y4P8
- Antigen synonyms:ATG18B|Atg21|CGI 50|DKFZp434J154|DKFZp686P02188|FLJ12979|FLJ14217|FLJ42984|WD repeat domain phosphoinositide-interacting protein 2|WD repeat domain|phosphoinositide interacting 2|WD40 repeat protein interacting with phosphoinositides 2|WIPI 2|WIPI-2|WIPI2_HUMAN|WIPI49 like protein 2|WIPI49-like protein 2
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 1:200, ELISA: 1:10000
Type:
Antigen: WIPI2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat