Anti-PON2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: PON2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77822-121
- 77822-120
- Antigen symbol:
- PON2
- PON2
- Antigen name:
- Serum paraoxonase/arylesterase 2 protein (32-97AA)
- Serum paraoxonase/arylesterase 2 protein (32-97AA)
- Antigen synonyms:
- A esterase 2|A-esterase 2|Aromatic esterase 2|Paraoxonase 2|Paraoxonase nirs|PON 2|PON2|PON2_HUMAN|Serum aryldialkylphosphatase 2|Serum paraoxonase/arylesterase 2
- A esterase 2|A-esterase 2|Aromatic esterase 2|Paraoxonase 2|Paraoxonase nirs|PON 2|PON2|PON2_HUMAN|Serum aryldialkylphosphatase 2|Serum paraoxonase/arylesterase 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15165
- Q15165
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together