Anti-ATXN7L3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ATXN7L3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-088
- 77822-087
- Antigen symbol:
- ATXN7L3
- ATXN7L3
- Antigen name:
- Ataxin-7-like protein 3 protein (51-347AA)
- Ataxin-7-like protein 3 protein (51-347AA)
- Antigen synonyms:
- AT7L3_HUMAN|Ataxin 7 like 3|Ataxin 7 like protein 3|Ataxin-7-like protein 3|atxn7l3|SAGA associated factor 11 homolog|SAGA-associated factor 11 homolog
- AT7L3_HUMAN|Ataxin 7 like 3|Ataxin 7 like protein 3|Ataxin-7-like protein 3|atxn7l3|SAGA associated factor 11 homolog|SAGA-associated factor 11 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14CW9
- Q14CW9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together