Anti-UGDH Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UGDH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-957
- 77803-957
- Antigen symbol:
- UGDH
- UGDH
- Antigen name:
- UDP-glucose 6-dehydrogenase protein (1-494AA)
- UDP-glucose 6-dehydrogenase protein (1-494AA)
- Antigen synonyms:
- GDH|UDP Glc dehydrogenase|UDP GlcDH|UDP glucose 6 dehydrogenase|UDP glucose dehydrogenase|UDP-Glc dehydrogenase|UDP-GlcDH|UDP-glucose 6-dehydrogenase|UDP-glucose dehydrogenase|UDPGDH|UGD|UGDH_HUMAN|Uridine diphospho glucose dehydrogenase
- GDH|UDP Glc dehydrogenase|UDP GlcDH|UDP glucose 6 dehydrogenase|UDP glucose dehydrogenase|UDP-Glc dehydrogenase|UDP-GlcDH|UDP-glucose 6-dehydrogenase|UDP-glucose dehydrogenase|UDPGDH|UGD|UGDH_HUMAN|Uridine diphospho glucose dehydrogenase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60701
- O60701
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together