Anti-TRMT1 Rabbit Polyclonal Antibody
Catalog # 77811-559
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:tRNA(m(2
- Antigen symbol:TRMT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NXH9
- Antigen synonyms:2)G26)dimethyltransferase|2-dimethylguanosine-26 methyltransferase|FLJ20244|N(2) N(2) dimethylguanosine tRNA methyltransferase|N(2)-N(2)) methyltransferase|TRM 1|TRM1 tRNA methyltransferase 1 homolog|TRM1_HUMAN|TRMT 1|Trmt1|tRNA (guanine(26)-N(2))-dimethyltransferase|tRNA 2 2 dimethylguanosine 26 methyltransferase|tRNA 2|tRNA(guanine 26 N(2) N(2)) methyltransferase|tRNA(guanine-26|tRNA(m(2 2)G26)dimethyltransferase|tRNA(m(2
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TRMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat