Anti-BIRC2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: BIRC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-443
- 77823-444
- Antigen symbol:
- BIRC2
- BIRC2
- Antigen name:
- Baculoviral IAP repeat-containing protein 2 protein (1-300AA)
- Baculoviral IAP repeat-containing protein 2 protein (1-300AA)
- Antigen synonyms:
- API 1|API1|Apoptosis inhibitor 1|Baculoviral IAP repeat containing 2|Baculoviral IAP repeat containing protein 2|Baculoviral IAP repeat-containing protein 2|BIRC 2|BIRC2|BIRC2_HUMAN|C IAP1|C-IAP1|Cellular inhibitor of apoptosis 1|cellular inhibitor of apoptosis protein 1|cIAP 1|cIAP1|HIAP 2|HIAP-2|HIAP2|IAP 2|IAP homolog B|IAP-2|IAP2|Inhibitor of apoptosis protein 2|MIHB|NFR2 TRAF signalling complex protein|RING finger protein 48|RNF 48|RNF48|TNFR2 TRAF signaling complex protein 2|TNFR2-TRAF-signaling complex protein 2
- API 1|API1|Apoptosis inhibitor 1|Baculoviral IAP repeat containing 2|Baculoviral IAP repeat containing protein 2|Baculoviral IAP repeat-containing protein 2|BIRC 2|BIRC2|BIRC2_HUMAN|C IAP1|C-IAP1|Cellular inhibitor of apoptosis 1|cellular inhibitor of apoptosis protein 1|cIAP 1|cIAP1|HIAP 2|HIAP-2|HIAP2|IAP 2|IAP homolog B|IAP-2|IAP2|Inhibitor of apoptosis protein 2|MIHB|NFR2 TRAF signalling complex protein|RING finger protein 48|RNF 48|RNF48|TNFR2 TRAF signaling complex protein 2|TNFR2-TRAF-signaling complex protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13490
- Q13490
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together