Anti-RLIM Rabbit Polyclonal Antibody
Catalog # 77797-379
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:RNF12_HUMAN
- Antigen symbol:RLIM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NVW2
- Antigen synonyms:E3 ubiquitin-protein ligase RLIM|LIM domain-interacting RING finger protein|R-LIM|Renal carcinoma antigen NY-REN-43|RING finger LIM domain-binding protein|RING finger protein 12|RLIM|RNF12|RNF12_HUMAN
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RLIM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat