Anti-GALE Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GALE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-011
- 77822-010
- Antigen symbol:
- GALE
- GALE
- Antigen name:
- UDP-glucose 4-epimerase protein (1-348AA)
- UDP-glucose 4-epimerase protein (1-348AA)
- Antigen synonyms:
- FLJ95174| FLJ97302| Galactose 4 epimerase UDP| Galactowaldenase| galE| GALE_HUMAN| OTTHUMP00000002991| OTTHUMP00000002994| OTTHUMP00000037931| OTTHUMP00000044857| SDR1E1| short chain dehydrogenase/reductase family 1E member 1| UDP galactose 4 epimerase| UDP galactose 4' epimerase| UDP glucose 4 epimerase| UDP-galactose 4-epimerase| UDP-glucose 4-epimerase
- FLJ95174| FLJ97302| Galactose 4 epimerase UDP| Galactowaldenase| galE| GALE_HUMAN| OTTHUMP00000002991| OTTHUMP00000002994| OTTHUMP00000037931| OTTHUMP00000044857| SDR1E1| short chain dehydrogenase/reductase family 1E member 1| UDP galactose 4 epimerase| UDP galactose 4' epimerase| UDP glucose 4 epimerase| UDP-galactose 4-epimerase| UDP-glucose 4-epimerase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14376
- Q14376
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together