Anti-NFIX Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NFIX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-115
- 77823-114
- Antigen symbol:
- NFIX
- NFIX
- Antigen name:
- Nuclear factor 1 X-type protein (201-440AA)
- Nuclear factor 1 X-type protein (201-440AA)
- Antigen synonyms:
- CCAAT box binding transcription factor|CCAAT-box-binding transcription factor|CTF|NF-I/X|NF1-X|NF1A|NF1X|NFI X|NFI-X|NFI/X|NFIX|NFIX_HUMAN|Nuclear factor 1 X type|Nuclear factor 1 X-type|Nuclear factor 1/X|Nuclear factor I/X|TGGCA binding protein|TGGCA-binding protein
- CCAAT box binding transcription factor|CCAAT-box-binding transcription factor|CTF|NF-I/X|NF1-X|NF1A|NF1X|NFI X|NFI-X|NFI/X|NFIX|NFIX_HUMAN|Nuclear factor 1 X type|Nuclear factor 1 X-type|Nuclear factor 1/X|Nuclear factor I/X|TGGCA binding protein|TGGCA-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14938
- Q14938
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together