Anti-FLII Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FLII
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-828
- 77822-829
- Antigen symbol:
- FLII
- FLII
- Antigen name:
- Protein flightless-1 homolog protein (577-729AA)
- Protein flightless-1 homolog protein (577-729AA)
- Antigen synonyms:
- Fli 1|FLI|Fli1|Flightless 1|Flightless I (Drosophila) homolog|Flightless I homolog|Flightless I homolog (Drosophila)|Flightless1|FlightlessI|fliI|FLII_HUMAN|Fliih|FLIL|MGC39265|Protein flightless 1 homolog|Protein flightless-1 homolog
- Fli 1|FLI|Fli1|Flightless 1|Flightless I (Drosophila) homolog|Flightless I homolog|Flightless I homolog (Drosophila)|Flightless1|FlightlessI|fliI|FLII_HUMAN|Fliih|FLIL|MGC39265|Protein flightless 1 homolog|Protein flightless-1 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13045
- Q13045
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together