Anti-PYC1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PYC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Baker\\\'s yeast
Specifications
- Catalog No:
- 77815-174
- 77815-173
- Antigen symbol:
- PYC1
- PYC1
- Antigen name:
- Recombinant Baker\\\'s yeast Pyruvate carboxylase 1 protein (18-470AA)
- Recombinant Baker\\\'s yeast Pyruvate carboxylase 1 protein (18-470AA)
- Antigen synonyms:
- PYC1|PYV|YGL062W|Pyruvate carboxylase 1|EC 6.4.1.1|Pyruvic carboxylase 1|PCB 1
- PYC1|PYV|YGL062W|Pyruvate carboxylase 1|EC 6.4.1.1|Pyruvic carboxylase 1|PCB 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Yeast
- Yeast
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11154
- P11154
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together