Anti-TRIM31 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: TRIM31
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-848
- 77841-849
- Antigen symbol:
- TRIM31
- TRIM31
- Antigen name:
- E3 ubiquitin-protein ligase TRIM31 protein (186-425AA)
- E3 ubiquitin-protein ligase TRIM31 protein (186-425AA)
- Antigen synonyms:
- C6orf13|RNF|E3 ubiquitin-protein ligase EC 2.3.2.27|RING-type E3 ubiquitin transferase Tripartite motif-containing protein 31
- C6orf13|RNF|E3 ubiquitin-protein ligase EC 2.3.2.27|RING-type E3 ubiquitin transferase Tripartite motif-containing protein 31
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BZY9
- Q9BZY9
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together