Anti-NOP10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NOP10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-635
- 77838-634
- Antigen symbol:
- NOP10
- NOP10
- Antigen name:
- H/ACA ribonucleoprotein complex subunit 3 protein (1-64AA)
- H/ACA ribonucleoprotein complex subunit 3 protein (1-64AA)
- Antigen synonyms:
- DKCB1| H/ACA ribonucleoprotein complex subunit 3| homolog of yeast Nop10p| MGC70651| NOLA3| NOP10| NOP10 ribonucleoprotein homolog (yeast)| NOP10_HUMAN| NOP10P| Nucleolar protein 10| Nucleolar protein family A member 3| nucleolar protein family A, member 3 (H/ACA small nucleolar RNPs)| snoRNP protein NOP10
- DKCB1| H/ACA ribonucleoprotein complex subunit 3| homolog of yeast Nop10p| MGC70651| NOLA3| NOP10| NOP10 ribonucleoprotein homolog (yeast)| NOP10_HUMAN| NOP10P| Nucleolar protein 10| Nucleolar protein family A member 3| nucleolar protein family A, member 3 (H/ACA small nucleolar RNPs)| snoRNP protein NOP10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPE3
- Q9NPE3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together