Anti-SMAD1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SMAD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-720
- 77823-719
- Antigen symbol:
- SMAD1
- SMAD1
- Antigen name:
- Mothers against decapentaplegic homolog 1 protein (150-250AA)
- Mothers against decapentaplegic homolog 1 protein (150-250AA)
- Antigen synonyms:
- BSP-1| BSP1| HsMAD1| JV4-1| JV41| MAD homolog 1| MAD mothers against decapentaplegic homolog 1| Mad related protein 1| Mad-related protein 1| MADH1| MADR1| Mothers against decapentaplegic homolog 1| Mothers against DPP homolog 1| SMA- AND MAD-RELATED PROTEIN 1| SMAD 1| SMAD family member 1| SMAD mothers against DPP homolog 1| Smad1| SMAD1_HUMAN| TGF beta signaling protein 1| Transforming growth factor-beta-signaling protein 1
- BSP-1| BSP1| HsMAD1| JV4-1| JV41| MAD homolog 1| MAD mothers against decapentaplegic homolog 1| Mad related protein 1| Mad-related protein 1| MADH1| MADR1| Mothers against decapentaplegic homolog 1| Mothers against DPP homolog 1| SMA- AND MAD-RELATED PROTEIN 1| SMAD 1| SMAD family member 1| SMAD mothers against DPP homolog 1| Smad1| SMAD1_HUMAN| TGF beta signaling protein 1| Transforming growth factor-beta-signaling protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15797
- Q15797
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together