Anti-FILIP1L Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FILIP1L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-666
- 77826-667
- Antigen symbol:
- FILIP1L
- FILIP1L
- Antigen name:
- Filamin A-interacting protein 1-like protein (1-200AA)
- Filamin A-interacting protein 1-like protein (1-200AA)
- Antigen synonyms:
- 130 kDa GPBP-interacting protein|90 kDa GPBP-interacting protein|COL4A3BPIP|DOC-1|DOC1|Downregulated in ovarian cancer 1 81 kDa isoform|FIL1L_HUMAN|Filamin A-interacting protein 1-like|FILIP1L|GIP130|GIP130a|GIP130b|GIP130c|GIP90|GPBP interacting protein 90|Protein down-regulated in ovarian cancer 1
- 130 kDa GPBP-interacting protein|90 kDa GPBP-interacting protein|COL4A3BPIP|DOC-1|DOC1|Downregulated in ovarian cancer 1 81 kDa isoform|FIL1L_HUMAN|Filamin A-interacting protein 1-like|FILIP1L|GIP130|GIP130a|GIP130b|GIP130c|GIP90|GPBP interacting protein 90|Protein down-regulated in ovarian cancer 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q4L180
- Q4L180
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together