Anti-DRAP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DRAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77808-435
- 77808-434
- Antigen symbol:
- DRAP1
- DRAP1
- Antigen name:
- Dr1-associated corepressor protein (4-198AA)
- Dr1-associated corepressor protein (4-198AA)
- Antigen synonyms:
- DR1 associated corepressor|DR1 associated protein 1 (negative cofactor 2 alpha)|DR1 associated protein 1|DR1 associated protein1|Dr1-associated corepressor|Dr1-associated protein 1|DRAP 1|Drap1|NC 2 alpha|NC 2alpha|NC2 alpha|NC2-alpha|NC2A_HUMAN|Negative co-factor 2-alpha|Negative cofactor 2 alpha|Negative cofactor 2alpha
- DR1 associated corepressor|DR1 associated protein 1 (negative cofactor 2 alpha)|DR1 associated protein 1|DR1 associated protein1|Dr1-associated corepressor|Dr1-associated protein 1|DRAP 1|Drap1|NC 2 alpha|NC 2alpha|NC2 alpha|NC2-alpha|NC2A_HUMAN|Negative co-factor 2-alpha|Negative cofactor 2 alpha|Negative cofactor 2alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14919
- Q14919
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together