Anti-Genome polyprotein Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Genome polyprotein
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hepatitis C virus genotype 1a
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77815-970
- 77815-969
- Antigen symbol:
- Genome polyprotein
- Genome polyprotein
- Antigen name:
- Recombinant Hepatitis C virus genotype 1a Genome polyprotein protein (192-325AA)
- Recombinant Hepatitis C virus genotype 1a Genome polyprotein protein (192-325AA)
- Antigen synonyms:
- Genome polyprotein [Cleaved into: Core protein p21| Capsid protein C| p21)| Core protein p19| Envelope glycoprotein E1| gp32| gp35)| Envelope glycoprotein E2| NS1| gp68| gp70)| p7| Protease NS2-3| p23| EC 3.4.22.-)| Serine protease NS3| EC 3.4.21.98| EC 3.6.1.15| EC 3.6.4.13| Hepacivirin| NS3P| p70)| Non-structural protein 4A| NS4A| p8)| Non-structural protein 4B| NS4B| p27)| Non-structural protein 5A| NS5A| p56)| RNA-directed RNA polymerase| EC 2.7.7.48| NS5B| p68)]
- Genome polyprotein [Cleaved into: Core protein p21| Capsid protein C| p21)| Core protein p19| Envelope glycoprotein E1| gp32| gp35)| Envelope glycoprotein E2| NS1| gp68| gp70)| p7| Protease NS2-3| p23| EC 3.4.22.-)| Serine protease NS3| EC 3.4.21.98| EC 3.6.1.15| EC 3.6.4.13| Hepacivirin| NS3P| p70)| Non-structural protein 4A| NS4A| p8)| Non-structural protein 4B| NS4B| p27)| Non-structural protein 5A| NS5A| p56)| RNA-directed RNA polymerase| EC 2.7.7.48| NS5B| p68)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Hepatitis C virus genotype 1a
- Hepatitis C virus genotype 1a
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P26664
- P26664
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together