Anti-SYF2 Rabbit Polyclonal Antibody
Catalog # 77801-680
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:SYF2_HUMAN
- Antigen symbol:SYF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O95926
- Antigen synonyms:CBPIN|CCNDBP1 interactor|CCNDBP1-interactor|DKFZp564O2082|fSAP29|Functional spliceosome associated protein 29|GCIP interacting protein p29|GCIPIP|NTC31|P29|Pre mRNA splicing factor SYF2|Pre-mRNA-splicing factor syf2|syf2|SYF2 homolog|SYF2 homolog RNA splicing factor (S. cerevisiae)|SYF2 homolog RNA splicing factor|SYF2_HUMAN
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SYF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat